Product Details
- SNP ID
-
rs148161858
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:26087508 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCAGACCGCGGTCCTGCAGGGGC[A/G]CTTGCTGCGTGAGTCCGAGGGCTGC
- Phenotype
-
MIM: 613609
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HFE
PubMed Links
Gene Details
- Gene
- HFE
- Gene Name
- hemochromatosis
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000410.3 |
228 |
Missense Mutation |
CAC,CGC |
H23R |
NP_000401.1 |
NM_001300749.1 |
228 |
Missense Mutation |
CAC,CGC |
H23R |
NP_001287678.1 |
NM_139003.2 |
228 |
Missense Mutation |
CAC,CGC |
H23R |
NP_620572.1 |
NM_139004.2 |
228 |
Missense Mutation |
CAC,CGC |
H23R |
NP_620573.1 |
NM_139006.2 |
228 |
Missense Mutation |
CAC,CGC |
H23R |
NP_620575.1 |
NM_139007.2 |
228 |
Missense Mutation |
CAC,CGC |
H23R |
NP_620576.1 |
NM_139008.2 |
228 |
Missense Mutation |
CAC,CGC |
H23R |
NP_620577.1 |
NM_139009.2 |
228 |
Missense Mutation |
CAC,CGC |
H23R |
NP_620578.1 |
NM_139010.2 |
228 |
Missense Mutation |
CAC,CGC |
H23R |
NP_620579.1 |
NM_139011.2 |
228 |
Missense Mutation |
CAC,CGC |
H23R |
NP_620580.1 |
XM_011514543.2 |
228 |
Missense Mutation |
CAC,CGC |
H23R |
XP_011512845.1 |
View Full Product Details