Product Details

SNP ID
rs148341961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27451852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTATGCTGAATAAGGGATGAACC[A/G]TAACTGAAGGTTTTCCCACATTCAT
Phenotype
MIM: 602277
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF184 PubMed Links

Gene Details

Gene
ZNF184
Gene Name
zinc finger protein 184
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318891.1 1992 Silent Mutation TAC,TAT Y569Y NP_001305820.1
NM_001318892.1 1992 Silent Mutation TAC,TAT Y569Y NP_001305821.1
NM_001318893.1 1992 Silent Mutation TAC,TAT Y493Y NP_001305822.1
NM_007149.2 1992 Silent Mutation TAC,TAT Y569Y NP_009080.2
XM_011514861.2 1992 Silent Mutation TAC,TAT Y493Y XP_011513163.1

View Full Product Details