Product Details

SNP ID
rs149031673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:53127222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGGGAAGTCAGTGAGGAAGAGG[C/G]AAGGAAACAAGAGGCAAGGAGACCA
Phenotype
MIM: 603715
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GCM1 PubMed Links

Gene Details

Gene
GCM1
Gene Name
glial cells missing homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003643.3 2504 UTR 3 NP_003634.2
XM_017011390.1 2504 UTR 3 XP_016866879.1

View Full Product Details