Product Details

SNP ID
rs150184697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29307154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTGCTGGGGGCCTCTCTGGGCT[C/T]ATGCATGCTGCCATTAACTTCTCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR14J1 PubMed Links

Gene Details

Gene
OR14J1
Gene Name
olfactory receptor family 14 subfamily J member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030946.1 465 Silent Mutation CTC,CTT L155L NP_112208.1

View Full Product Details