Product Details

SNP ID
rs150493791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:55174800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTGTTCGTCGTGGCTCTCATTGG[A/G]AACGTCCTGGGTGAGTCTCCTCCCG
Phenotype
MIM: 602393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCRTR2 PubMed Links

Gene Details

Gene
HCRTR2
Gene Name
hypocretin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001526.3 528 Silent Mutation GGA,GGG G71G NP_001517.2
XM_017010798.1 528 Silent Mutation GGA,GGG G71G XP_016866287.1

View Full Product Details