Product Details

SNP ID
rs151064538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26104067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCGGCTATTCGCCGTTTGGCTCG[A/G]CGCGGTGGCGTCAAGCGCATTTCCG
Phenotype
MIM: 613609 MIM: 142712 MIM: 602827
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HFE PubMed Links

Gene Details

Gene
HFE
Gene Name
hemochromatosis
There are no transcripts associated with this gene.

Gene
HIST1H1T
Gene Name
histone cluster 1, H1t
There are no transcripts associated with this gene.

Gene
HIST1H4C
Gene Name
histone cluster 1, H4c
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003542.3 120 Silent Mutation CGA,CGG R40R NP_003533.1

View Full Product Details