Product Details

SNP ID
rs138327413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:899037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCCGCCCGCCCCCCTCACCTGC[A/G]TACTCCTGGGGCAGCATGGGCCTGT
Phenotype
MIM: 608114 MIM: 612056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAP1 PubMed Links

Gene Details

Gene
ADAP1
Gene Name
ArfGAP with dual PH domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284308.1 1125 Silent Mutation TAC,TAT Y375Y NP_001271237.1
NM_001284309.1 1125 Silent Mutation TAC,TAT Y292Y NP_001271238.1
NM_001284310.1 1125 Silent Mutation TAC,TAT Y292Y NP_001271239.1
NM_001284311.1 1125 Silent Mutation TAC,TAT Y269Y NP_001271240.1
NM_006869.3 1125 Silent Mutation TAC,TAT Y364Y NP_006860.1
Gene
GET4
Gene Name
golgi to ER traffic protein 4
There are no transcripts associated with this gene.

View Full Product Details