Product Details

SNP ID
rs138625059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:138596471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAGTGAATGCAGAAATGGCGCA[C/T]ACAACACAGACAGATGAGAAGAGAC
Phenotype
MIM: 611700
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SVOPL PubMed Links

Gene Details

Gene
SVOPL
Gene Name
SVOP like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139456.1 1104 Silent Mutation GTA,GTG V471V NP_001132928.1
NM_174959.3 1104 Silent Mutation GTA,GTG V319V NP_777619.1
XM_005250143.3 1104 Silent Mutation GTA,GTG V471V XP_005250200.1
XM_011515797.2 1104 Silent Mutation GTA,GTG V319V XP_011514099.1
XM_017011746.1 1104 Silent Mutation GTA,GTG V380V XP_016867235.1
XM_017011747.1 1104 Silent Mutation GTA,GTG V319V XP_016867236.1
XM_017011748.1 1104 Silent Mutation GTA,GTG V319V XP_016867237.1

View Full Product Details