Product Details

SNP ID
rs139104239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107924291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGTTTTTGCTTCATTTTGTAG[C/G]ATTTCGGCTTTCCTTCTGGCATCAG
Phenotype
MIM: 238331 MIM: 150240
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DLD PubMed Links

Gene Details

Gene
DLD
Gene Name
dihydrolipoamide dehydrogenase
There are no transcripts associated with this gene.

Gene
LAMB1
Gene Name
laminin subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002291.2 5498 Missense Mutation ATC,ATG I1721M NP_002282.2
XM_017012201.1 5498 Missense Mutation ATC,ATG I1745M XP_016867690.1
XM_017012202.1 5498 Intron XP_016867691.1

View Full Product Details