Product Details

SNP ID
rs139452042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:900111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCGGCACACCCACCTTGGGCCC[C/T]GTCTTCTCCATGTAGCCTTCCTTCA
Phenotype
MIM: 608114 MIM: 612056
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAP1 PubMed Links

Gene Details

Gene
ADAP1
Gene Name
ArfGAP with dual PH domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284308.1 819 Silent Mutation ACC,ACT T273T NP_001271237.1
NM_001284309.1 819 Silent Mutation ACC,ACT T190T NP_001271238.1
NM_001284310.1 819 Silent Mutation ACC,ACT T190T NP_001271239.1
NM_001284311.1 819 Silent Mutation ACC,ACT T167T NP_001271240.1
NM_006869.3 819 Silent Mutation ACC,ACT T262T NP_006860.1
Gene
GET4
Gene Name
golgi to ER traffic protein 4
There are no transcripts associated with this gene.

View Full Product Details