Product Details

SNP ID
rs139740364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:7637894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGCTATCTTCTTTAAACTGGA[C/T]ATAAGATGTACACAAGATGGTGGCC
Phenotype
MIM: 179837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPA3 PubMed Links

Gene Details

Gene
RPA3
Gene Name
replication protein A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002947.4 1425 Missense Mutation ATC,GTC I85V NP_002938.1
Gene
UMAD1
Gene Name
UBAP1-MVB12-associated (UMA) domain containing 1
There are no transcripts associated with this gene.

View Full Product Details