Product Details
- SNP ID
-
rs140530900
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:51017565 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCTGTTCATTCACACGAGCAAGG[A/G]CACCTGCAGGGAAGAGAGATTCACA
- Phenotype
-
MIM: 610317
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
COBL
PubMed Links
Gene Details
- Gene
- COBL
- Gene Name
- cordon-bleu WH2 repeat protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001287436.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1268S |
NP_001274365.1 |
NM_001287438.1 |
3993 |
Intron |
|
|
NP_001274367.1 |
NM_015198.3 |
3993 |
Missense Mutation |
CCC,TCC |
P1258S |
NP_056013.2 |
XM_005271750.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1330S |
XP_005271807.1 |
XM_005271751.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1315S |
XP_005271808.1 |
XM_005271756.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1211S |
XP_005271813.1 |
XM_011515234.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1355S |
XP_011513536.1 |
XM_011515235.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1340S |
XP_011513537.1 |
XM_011515236.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1308S |
XP_011513538.1 |
XM_011515237.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1298S |
XP_011513539.1 |
XM_011515238.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1267S |
XP_011513540.1 |
XM_011515239.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1251S |
XP_011513541.1 |
XM_011515240.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1236S |
XP_011513542.1 |
XM_011515241.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1044S |
XP_011513543.1 |
XM_017011898.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1273S |
XP_016867387.1 |
XM_017011899.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1227S |
XP_016867388.1 |
XM_017011900.1 |
3993 |
Missense Mutation |
CCC,TCC |
P1170S |
XP_016867389.1 |
XM_017011901.1 |
3993 |
Intron |
|
|
XP_016867390.1 |
View Full Product Details