Product Details

SNP ID
rs141292389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:138594621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTTGCAGGTCTTCATTTAATTT[G/T]CTGGAAGAAAGTTATTTAATAGATC
Phenotype
MIM: 611700 MIM: 603406
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SVOPL PubMed Links

Gene Details

Gene
SVOPL
Gene Name
SVOP like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139456.1 1159 Missense Mutation AAA,CAA K490Q NP_001132928.1
NM_174959.3 1159 Missense Mutation AAA,CAA K338Q NP_777619.1
XM_005250143.3 1159 Missense Mutation AAA,CAA K490Q XP_005250200.1
XM_011515797.2 1159 Missense Mutation AAA,CAA K338Q XP_011514099.1
XM_017011746.1 1159 Missense Mutation AAA,CAA K399Q XP_016867235.1
XM_017011747.1 1159 Missense Mutation AAA,CAA K338Q XP_016867236.1
XM_017011748.1 1159 Missense Mutation AAA,CAA K338Q XP_016867237.1
Gene
TRIM24
Gene Name
tripartite motif containing 24
There are no transcripts associated with this gene.

View Full Product Details