Product Details

SNP ID
rs141398137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:138596443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGGCCCGTCCTTTGGTTTCGA[C/T]GGGGAGAGTGAATGCAGAAATGGCG
Phenotype
MIM: 611700
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SVOPL PubMed Links

Gene Details

Gene
SVOPL
Gene Name
SVOP like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139456.1 1132 Missense Mutation ATC,GTC I481V NP_001132928.1
NM_174959.3 1132 Missense Mutation ATC,GTC I329V NP_777619.1
XM_005250143.3 1132 Missense Mutation ATC,GTC I481V XP_005250200.1
XM_011515797.2 1132 Missense Mutation ATC,GTC I329V XP_011514099.1
XM_017011746.1 1132 Missense Mutation ATC,GTC I390V XP_016867235.1
XM_017011747.1 1132 Missense Mutation ATC,GTC I329V XP_016867236.1
XM_017011748.1 1132 Missense Mutation ATC,GTC I329V XP_016867237.1

View Full Product Details