Product Details
- SNP ID
-
rs143177014
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:5136653 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAGGGACCCACCTTGCTGACAAA[C/G]TCCGATCCTGAGCTTTTGCAAAGAC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF890P
PubMed Links
Gene Details
- Gene
- ZNF890P
- Gene Name
- zinc finger protein 890, pseudogene
There are no transcripts associated with this gene.
View Full Product Details