Product Details

SNP ID
rs145987368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:23182192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGAGGAGGACGGCAGCAACCGC[A/G]GCAGCAGCCTTCAGGTGACTCTCCT
Phenotype
MIM: 611119
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL7 PubMed Links

Gene Details

Gene
KLHL7
Gene Name
kelch like family member 7
There are no transcripts associated with this gene.

Gene
NUPL2
Gene Name
nucleoporin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007342.2 626 Missense Mutation CAG,CGG Q36R NP_031368.1
XM_005249592.1 626 Missense Mutation CAG,CGG Q36R XP_005249649.1
XM_005249593.3 626 UTR 5 XP_005249650.1
XM_011515104.2 626 UTR 5 XP_011513406.1
XM_017011714.1 626 UTR 5 XP_016867203.1
XM_017011715.1 626 UTR 5 XP_016867204.1
XM_017011716.1 626 UTR 5 XP_016867205.1
XM_017011717.1 626 UTR 5 XP_016867206.1
XM_017011718.1 626 Intron XP_016867207.1

View Full Product Details