Product Details

SNP ID
rs146017906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123452867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGCACTGGAACCTGAGAAAAA[C/T]AGTTCTTAGCCAGGATATGTTTGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQUB PubMed Links

Gene Details

Gene
IQUB
Gene Name
IQ motif and ubiquitin domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282855.1 2841 Missense Mutation TAT,TGT Y751C NP_001269784.1
NM_001321293.1 2841 Missense Mutation TAT,TGT Y751C NP_001308222.1
NM_178827.4 2841 Missense Mutation TAT,TGT Y751C NP_849149.3
XM_005250161.3 2841 Missense Mutation TAT,TGT Y751C XP_005250218.1
XM_005250162.4 2841 Intron XP_005250219.1
XM_011515833.2 2841 Missense Mutation TAT,TGT Y751C XP_011514135.1
XM_011515834.2 2841 Missense Mutation TAT,TGT Y751C XP_011514136.1
XM_017011771.1 2841 Missense Mutation TAT,TGT Y751C XP_016867260.1
XM_017011772.1 2841 Missense Mutation TAT,TGT Y472C XP_016867261.1

View Full Product Details