Product Details

SNP ID
rs146593231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103103521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTTAAGTCCGTATCTCTCTAGAG[C/T]TTCATTCAGCTTCACTGGAGGCTCT
Phenotype
MIM: 611864 MIM: 612334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARMC10 PubMed Links

Gene Details

Gene
ARMC10
Gene Name
armadillo repeat containing 10
There are no transcripts associated with this gene.

Gene
NAPEPLD
Gene Name
N-acyl phosphatidylethanolamine phospholipase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122838.1 1418 Missense Mutation ACT,GCT T364A NP_001116310.1
NM_198990.4 1418 Missense Mutation ACT,GCT T364A NP_945341.3
XM_005250214.4 1418 Missense Mutation ACT,GCT T364A XP_005250271.2
XM_005250215.2 1418 Missense Mutation ACT,GCT T364A XP_005250272.1
XM_005250218.2 1418 Missense Mutation ACT,GCT T364A XP_005250275.1
XM_011515933.2 1418 Missense Mutation ACT,GCT T364A XP_011514235.1
XM_011515934.2 1418 Missense Mutation ACT,GCT T229A XP_011514236.1
XM_017011854.1 1418 Missense Mutation ACT,GCT T364A XP_016867343.1
XM_017011855.1 1418 Missense Mutation ACT,GCT T364A XP_016867344.1
XM_017011856.1 1418 Intron XP_016867345.1
XM_017011857.1 1418 Intron XP_016867346.1

View Full Product Details