Product Details

SNP ID
rs147347140
Assay Type
Functionally Tested
NCBI dbSNP Submissions
6
Location
Chr.7:156950048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGGCTGCAGAGGACGGCGGGCC[C/T]CGAACAGGGTCCCGGCCTGGGAGGC
Phenotype
MIM: 611269
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NOM1 PubMed Links

Gene Details

Gene
NOM1
Gene Name
nucleolar protein with MIF4G domain 1
There are no transcripts associated with this gene.

View Full Product Details