Product Details

SNP ID
rs147474808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141837979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGAGCATGGCAGGCTTAGCCAGC[C/T]TGATGAGCATGAGGTCATCCTGTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS37 PubMed Links

Gene Details

Gene
PRSS37
Gene Name
protease, serine 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008270.2 1805 Missense Mutation AAG,AGG K104R NP_001008271.2
NM_001171951.1 1805 Missense Mutation AAG,AGG K104R NP_001165422.1
XM_005249946.4 1805 Missense Mutation AAG,AGG K104R XP_005250003.1
XM_005249947.3 1805 Missense Mutation AAG,AGG K104R XP_005250004.1
XM_011515792.2 1805 Missense Mutation AAG,AGG K41R XP_011514094.1
XM_011515793.2 1805 Missense Mutation AAG,AGG K41R XP_011514095.1

View Full Product Details