Product Details

SNP ID
rs147800282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101232451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCGTATTTGCCAAAGCTGCTGT[C/T]GCCTTCTTGGTCCGAGGTGGCGACG
Phenotype
MIM: 615778 MIM: 609003
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN15 PubMed Links

Gene Details

Gene
CLDN15
Gene Name
claudin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185080.1 1011 Missense Mutation AAC,GAC N216D NP_001172009.1
NM_014343.2 1011 Missense Mutation AAC,GAC N216D NP_055158.1
Gene
FIS1
Gene Name
fission, mitochondrial 1
There are no transcripts associated with this gene.

Gene
ZNHIT1
Gene Name
zinc finger HIT-type containing 1
There are no transcripts associated with this gene.

View Full Product Details