Product Details

SNP ID
rs148084694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130293208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTGGATACTGTTCATTGGGGCC[A/C]TTATTGGGTCCAGCATCTGTGGCCA
Phenotype
MIM: 600688 MIM: 607635
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CPA2 PubMed Links

Gene Details

Gene
CPA2
Gene Name
carboxypeptidase A2
There are no transcripts associated with this gene.

Gene
CPA4
Gene Name
carboxypeptidase A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163446.1 75 Missense Mutation ATT,CTT I10L NP_001156918.1
NM_016352.3 75 Missense Mutation ATT,CTT I10L NP_057436.2

View Full Product Details