Product Details

SNP ID
rs148985640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99400364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGTTGTGTGACCTTTTTGGTTG[A/T]CTGTGCCACCCGGTTGGGGTTCTCG
Phenotype
MIM: 604223 MIM: 603477 MIM: 607075
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARPC1B PubMed Links

Gene Details

Gene
ARPC1B
Gene Name
actin related protein 2/3 complex subunit 1B
There are no transcripts associated with this gene.

Gene
BUD31
Gene Name
BUD31 homolog
There are no transcripts associated with this gene.

Gene
PDAP1
Gene Name
PDGFA associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014891.6 408 Missense Mutation ACA,TCA T92S NP_055706.1

View Full Product Details