Product Details

SNP ID
rs150681972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19721659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAATTGTAGCATCCTTGAAGCC[A/G]TCTGTCTTTCAGAACAAAGACGTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM196 PubMed Links

Gene Details

Gene
TMEM196
Gene Name
transmembrane protein 196
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152774.3 1957 UTR 3 NP_689987.3
XM_005249692.4 1957 UTR 3 XP_005249749.2
XM_011515249.2 1957 UTR 3 XP_011513551.1
XM_011515250.2 1957 UTR 3 XP_011513552.1
XM_017011928.1 1957 Intron XP_016867417.1
XM_017011929.1 1957 Intron XP_016867418.1

View Full Product Details