Product Details
- SNP ID
-
rs150796882
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:75813407 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAACAAAGAACATGCAGCAGGGAGA[A/G]GGGATGACCACAGAGCCTAGAAGAG
- Phenotype
-
MIM: 602495
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCL24
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11465293,rs2302006] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCL24
- Gene Name
- C-C motif chemokine ligand 24
View Full Product Details