Product Details

SNP ID
rs151183604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149004120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTCCTTCTTGGGGTCCATCACAA[C/T]GGAGTCAAAGGTCTTTCCCACCACG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDIA4 PubMed Links

Gene Details

Gene
PDIA4
Gene Name
protein disulfide isomerase family A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004911.4 1894 Missense Mutation ATT,GTT I538V NP_004902.1
XM_006716185.2 1894 Missense Mutation ATT,GTT I539V XP_006716248.1

View Full Product Details