Product Details

SNP ID
rs151191365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107893213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTTTCTAGAGAGGCCATTTCA[A/G]TCGAATATCTCATGGCCTACAGGGA
Phenotype
MIM: 238331
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLD PubMed Links

Gene Details

Gene
DLD
Gene Name
dihydrolipoamide dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000108.4 197 Missense Mutation AAT,AGT N18S NP_000099.2
NM_001289750.1 197 UTR 5 NP_001276679.1
NM_001289751.1 197 Missense Mutation AAT,AGT N18S NP_001276680.1
NM_001289752.1 197 Missense Mutation AAT,AGT N18S NP_001276681.1

View Full Product Details