Product Details

SNP ID
rs117684375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:55452545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGTGCAGCCTCAATAGCTTGG[A/G]TTTCTAGATGGGCATCAGCCTCCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SBF1P1 PubMed Links

Gene Details

Gene
SBF1P1
Gene Name
SET binding factor 1 pseudogene 1
There are no transcripts associated with this gene.

Gene
XKR4
Gene Name
XK related 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052898.1 Intron NP_443130.1

View Full Product Details