Product Details

SNP ID
rs117889808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123015612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGCAGAAAGACGGGGACACAAA[C/T]GGAGAGAAGAGAACAAAGTCACTTC
Phenotype
MIM: 608813
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DERL1 PubMed Links

Gene Details

Gene
DERL1
Gene Name
derlin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134671.2 Intron NP_001128143.1
NM_024295.5 Intron NP_077271.1
XM_006716657.1 Intron XP_006716720.1
XM_011517299.1 Intron XP_011515601.1

View Full Product Details