Product Details

SNP ID
rs139157188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:99962441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAAACATTCACAAGAATGCTGTC[A/G]CTCTGGAAAAGACATGAAGTTTTAT
Phenotype
MIM: 615650
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RGS22 PubMed Links

Gene Details

Gene
RGS22
Gene Name
regulator of G-protein signaling 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005250860.2 Intron XP_005250917.1
XM_011516959.2 Intron XP_011515261.1

View Full Product Details