Product Details

SNP ID
rs140539975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143977999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGTGGTACAGCACCTGCTCCACC[A/G]GGCGCCGCTCGCATCGCTGCAGCAG
Phenotype
MIM: 609564 MIM: 601282
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARP10 PubMed Links

Gene Details

Gene
PARP10
Gene Name
poly(ADP-ribose) polymerase family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317895.1 2751 Missense Mutation CCG,CTG P892L NP_001304824.1
NM_032789.4 2751 Missense Mutation CCG,CTG P880L NP_116178.2
XM_011517336.2 2751 Missense Mutation CCG,CTG P871L XP_011515638.1
Gene
PLEC
Gene Name
plectin
There are no transcripts associated with this gene.

View Full Product Details