Product Details
- SNP ID
-
rs140675697
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:70644198 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAGATAGCATTTTCCTCTTCTAA[C/G]AGTAGAGCCATGTGATCATCAGTGT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LACTB2
PubMed Links
Gene Details
- Gene
- LACTB2
- Gene Name
- lactamase beta 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016027.2 |
551 |
Silent Mutation |
CTC,CTG |
L153L |
NP_057111.1 |
- Gene
- LACTB2-AS1
- Gene Name
- LACTB2 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details