Product Details

SNP ID
rs141975370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123015580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCTCTCCTACTGGGCAGCCAG[C/T]GGTACCTGGTGCAGAAAGACGGGGA
Phenotype
MIM: 608813
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DERL1 PubMed Links

Gene Details

Gene
DERL1
Gene Name
derlin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134671.2 724 Missense Mutation CAC,CGC H188R NP_001128143.1
NM_024295.5 724 Missense Mutation CAC,CGC H208R NP_077271.1
XM_006716657.1 724 Missense Mutation CAC,CGC H108R XP_006716720.1
XM_011517299.1 724 Missense Mutation CAC,CGC H108R XP_011515601.1

View Full Product Details