Product Details

SNP ID
rs141982652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23681368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCTTCTCCAAGTCTCCCAGCTC[C/G]GAGGAGAGCTGCTTTCGCTTAGTCT
Phenotype
MIM: 602041
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NKX3-1 PubMed Links

Gene Details

Gene
NKX3-1
Gene Name
NK3 homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256339.1 606 Silent Mutation TCC,TCG S111S NP_001243268.1
NM_006167.3 606 Silent Mutation TCC,TCG S186S NP_006158.2

View Full Product Details