Product Details
- SNP ID
-
rs142161778
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:17300169 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGAGTGCTTGCTGGGCTCACTT[G/T]GCTTACTCTTCACCTTAGTGCAATT
- Phenotype
-
MIM: 603562
MIM: 609927
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MTMR7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3764796] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MTMR7
- Gene Name
- myotubularin related protein 7
- Gene
- VPS37A
- Gene Name
- VPS37A, ESCRT-I subunit
View Full Product Details