Product Details
- SNP ID
-
rs145797598
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:121614461 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGTATAACACTGAGTAGAGAGAC[A/G]TGAAGACCATGACGATATTTCCTCT
- Phenotype
-
MIM: 601636
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HAS2
PubMed Links
Gene Details
- Gene
- HAS2
- Gene Name
- hyaluronan synthase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005328.2 |
1845 |
Missense Mutation |
ACG,ATG |
T436M |
NP_005319.1 |
View Full Product Details