Product Details

SNP ID
rs146349792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:97276664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGACCTTGCCTGCCTTCTTTT[C/T]CCTTTCCTTTCTCTACACGAGCCCC
Phenotype
MIM: 614721
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927066 PubMed Links

Gene Details

Gene
LOC101927066
Gene Name
uncharacterized LOC101927066
There are no transcripts associated with this gene.

Gene
TSPYL5
Gene Name
TSPY like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033512.2 1285 Missense Mutation GAA,GGA E394G NP_277047.2

View Full Product Details