Product Details

SNP ID
rs146436573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:120445509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCTGGTGATCTGGGGGGAAGG[A/C]AAATTCCCGTCGGCGGCCAGTAGGG
Phenotype
MIM: 610200 MIM: 605927
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MRPL13 PubMed Links

Gene Details

Gene
MRPL13
Gene Name
mitochondrial ribosomal protein L13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014078.5 109 Intron NP_054797.2
Gene
MTBP
Gene Name
MDM2 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022045.4 109 Silent Mutation GGA,GGC G13G NP_071328.2
XM_011516962.2 109 Silent Mutation GGA,GGC G13G XP_011515264.1
XM_011516963.2 109 Silent Mutation GGA,GGC G13G XP_011515265.1

View Full Product Details