Product Details

SNP ID
rs146849251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86048697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAGGGCCTGGCCAGCTGACATG[C/T]GATGACCAGCCTCCAAAATCAGTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSKH2 PubMed Links

Gene Details

Gene
PSKH2
Gene Name
protein serine kinase H2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033126.1 923 Missense Mutation CAC,CGC H308R NP_149117.1
XM_017013929.1 923 Missense Mutation CAC,CGC H427R XP_016869418.1

View Full Product Details