Product Details

SNP ID
rs148135759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144881698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCATATGGCTTCTCGCCCGTGTG[G/T]GTCCTCAGGTGCACAATCAGAGTTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF250 PubMed Links

Gene Details

Gene
ZNF250
Gene Name
zinc finger protein 250
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109689.3 1617 Silent Mutation ACA,ACC T495T NP_001103159.1
NM_021061.4 1617 Silent Mutation ACA,ACC T500T NP_066405.1
XM_005272327.1 1617 Silent Mutation ACA,ACC T495T XP_005272384.1
XM_005272328.3 1617 Silent Mutation ACA,ACC T495T XP_005272385.1
XM_006716612.3 1617 Silent Mutation ACA,ACC T500T XP_006716675.1
XM_006716613.3 1617 Silent Mutation ACA,ACC T500T XP_006716676.1
XM_011517209.1 1617 Silent Mutation ACA,ACC T495T XP_011515511.1

View Full Product Details