Product Details

SNP ID
rs148436633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:26386237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATTAAAAATTACAGGGCAAAT[A/C]CCCAAACTCCTTCCAAGTCAAAAGG
Phenotype
MIM: 605368
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BNIP3L PubMed Links

Gene Details

Gene
BNIP3L
Gene Name
BCL2 interacting protein 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004331.2 Intron NP_004322.1
XM_005273617.3 Intron XP_005273674.1
Gene
SDAD1P1
Gene Name
SDA1 domain containing 1 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details