Product Details

SNP ID
rs148952091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:103401985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATTGATATGCTGGTTGTACTTCA[A/T]CTTCAGCAATTCATATGCCATAAAC
Phenotype
MIM: 610815
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A32 PubMed Links

Gene Details

Gene
SLC25A32
Gene Name
solute carrier family 25 member 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030780.4 1020 Missense Mutation ATG,TTG M208L NP_110407.2
XM_017013877.1 1020 Missense Mutation ATG,TTG M76L XP_016869366.1

View Full Product Details