Product Details

SNP ID
rs138071426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136334504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCGAGCGTCTGTGCAAGGCGGG[C/T]GACTTCAAGACAGGCGTGGCCTTCT
Phenotype
MIM: 609491
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DKFZP434A062 PubMed Links

Gene Details

Gene
DKFZP434A062
Gene Name
uncharacterized LOC26102
There are no transcripts associated with this gene.

Gene
GPSM1
Gene Name
G-protein signaling modulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145638.2 346 Silent Mutation GGC,GGT G42G NP_001139110.2
NM_001145639.1 346 Intron NP_001139111.1
NM_001200003.1 346 Intron NP_001186932.1
NM_015597.5 346 Silent Mutation GGC,GGT G42G NP_056412.5
XM_011518498.2 346 Silent Mutation GGC,GGT G74G XP_011516800.1
XM_017014598.1 346 Silent Mutation GGC,GGT G19G XP_016870087.1
XM_017014599.1 346 Intron XP_016870088.1

View Full Product Details