Product Details

SNP ID
rs138244015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92618867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTAGAAGAATGTGGAACATTCC[A/G]CAAATACTGGGTGCAGGGTTTAGCA
Phenotype
MIM: 611505
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPP PubMed Links

Gene Details

Gene
CENPP
Gene Name
centromere protein P
There are no transcripts associated with this gene.

Gene
IPPK
Gene Name
inositol-pentakisphosphate 2-kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022755.5 Intron NP_073592.1
XM_017015041.1 Intron XP_016870530.1
Gene
LOC100128361
Gene Name
uncharacterized LOC100128361
There are no transcripts associated with this gene.

View Full Product Details