Product Details

SNP ID
rs138701847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:108934503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTCGCCATGGCTACCGAGCCCG[A/C]AGCCGCGGAGCCGGTGGTGCCTTCG
Phenotype
MIM: 604785 MIM: 603722
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CTNNAL1 PubMed Links

Gene Details

Gene
CTNNAL1
Gene Name
catenin alpha like 1
There are no transcripts associated with this gene.

Gene
FAM206A
Gene Name
family with sequence similarity 206 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017832.3 107 Missense Mutation GAA,GCA E6A NP_060302.1
XM_011518816.2 107 Missense Mutation GAA,GCA E6A XP_011517118.1
Gene
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318360.1 107 Intron NP_001305289.1
NM_003640.4 107 Intron NP_003631.2
XM_011519136.1 107 Intron XP_011517438.1
XM_011519137.1 107 Intron XP_011517439.1
XM_017015238.1 107 Intron XP_016870727.1

View Full Product Details