Product Details

SNP ID
rs138792663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120858088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGTGATAGATGACTTGAGGTG[C/G]GACAGGGATGAGTCATCTTCACTGA
Phenotype
MIM: 609740
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009936.2 1723 Intron NP_001009936.1
NM_001286840.1 1723 Silent Mutation TCC,TCG S552S NP_001273769.1
NM_001286842.1 1723 Silent Mutation TCC,TCG S324S NP_001273771.1
NM_001286843.1 1723 Intron NP_001273772.1
NM_015651.2 1723 Silent Mutation TCC,TCG S533S NP_056466.1
XM_005251906.2 1723 Silent Mutation TCC,TCG S533S XP_005251963.1
XM_011518509.2 1723 Silent Mutation TCC,TCG S533S XP_011516811.1
XM_011518511.2 1723 Intron XP_011516813.1
XM_011518515.2 1723 Intron XP_011516817.1
XM_011518516.2 1723 Intron XP_011516818.1
XM_017014612.1 1723 Silent Mutation TCC,TCG S533S XP_016870101.1
XM_017014613.1 1723 Intron XP_016870102.1
Gene
PSMD5-AS1
Gene Name
PSMD5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details