Product Details

SNP ID
rs139595138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37425959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCATCAGCACCATGTCTGTGGG[A/C]ATCGACCACTTGGCTTTGGATGAAA
Phenotype
MIM: 604296
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GRHPR PubMed Links

Gene Details

Gene
GRHPR
Gene Name
glyoxylate reductase/hydroxypyruvate reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012203.1 307 Silent Mutation GGA,GGC G84G NP_036335.1
XM_005251631.1 307 Intron XP_005251688.1
XM_011518073.2 307 UTR 5 XP_011516375.1
XM_017015320.1 307 Silent Mutation GGA,GGC G84G XP_016870809.1
XM_017015321.1 307 Silent Mutation GGA,GGC G84G XP_016870810.1
XM_017015322.1 307 Intron XP_016870811.1
XM_017015323.1 307 UTR 5 XP_016870812.1

View Full Product Details