Product Details
- SNP ID
-
rs142090142
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:128122478 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGCGCACGTTGTCCTGGAGGCGGT[A/G]CCTGGGCGGGGAAGGGAAAAGCCCC
- Phenotype
-
MIM: 608152
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PTGES2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs13283456] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PTGES2
- Gene Name
- prostaglandin E synthase 2
- Gene
- PTGES2-AS1
- Gene Name
- PTGES2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
- Gene
- SLC25A25-AS1
- Gene Name
- SLC25A25 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details