Product Details

SNP ID
rs143856051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128522679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAACCTTTTCAGGAGGGCCTGAG[A/G]CTATGGCAGGAGGAGCAGGAGAGGA
Phenotype
MIM: 603371
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLE1 PubMed Links

Gene Details

Gene
GLE1
Gene Name
GLE1, RNA export mediator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003722.1 670 Silent Mutation AGA,AGG R148R NP_001003722.1
NM_001499.2 670 Silent Mutation AGA,AGG R148R NP_001490.1
XM_006717059.2 670 Silent Mutation AGA,AGG R151R XP_006717122.1
XM_006717060.2 670 Silent Mutation AGA,AGG R151R XP_006717123.1
XM_011518549.1 670 Silent Mutation AGA,AGG R151R XP_011516851.1
XM_011518550.1 670 Silent Mutation AGA,AGG R151R XP_011516852.1
XM_011518551.1 670 Silent Mutation AGA,AGG R148R XP_011516853.1

View Full Product Details