Product Details

SNP ID
rs144405933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120858197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCACTGCTGTCCGAGGGGCAG[C/T]GTCCATCCAGCTCAGCTGCCCACCG
Phenotype
MIM: 609740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009936.2 1614 Intron NP_001009936.1
NM_001286840.1 1614 Missense Mutation CAC,CGC H516R NP_001273769.1
NM_001286842.1 1614 Missense Mutation CAC,CGC H288R NP_001273771.1
NM_001286843.1 1614 Intron NP_001273772.1
NM_015651.2 1614 Missense Mutation CAC,CGC H497R NP_056466.1
XM_005251906.2 1614 Missense Mutation CAC,CGC H497R XP_005251963.1
XM_011518509.2 1614 Missense Mutation CAC,CGC H497R XP_011516811.1
XM_011518511.2 1614 UTR 3 XP_011516813.1
XM_011518515.2 1614 Intron XP_011516817.1
XM_011518516.2 1614 Intron XP_011516818.1
XM_017014612.1 1614 Missense Mutation CAC,CGC H497R XP_016870101.1
XM_017014613.1 1614 Intron XP_016870102.1
Gene
PSMD5-AS1
Gene Name
PSMD5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details